Thermo Scientific™

CEDIA™ Therapeutic Drug Monitoring (TDM) Calibrators

Catalog number: 100017
Thermo Scientific™

CEDIA™ Therapeutic Drug Monitoring (TDM) Calibrators

Catalog number: 100017
Streamline testing with Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Calibrators, a single calibrator kit that can be used with multiple CEDIA TDM assays. These economical kits are valued for their accuracy, precision and lot-to-lot dependability. They are available for cardiac or antiarrhythmic, antiepileptic and antibiotic drug monitoring in human serum and plasma. Each kit includes both high and low calibrators.
 
Catalog Number
100017
Unit Size
Each
Description
CEDIA Antibiotic TDM Multi-Calibrators
Detectable Analytes
Gentamicin
Price (USD)
Full specifications
DescriptionCEDIA Antibiotic TDM Multi-Calibrators
Detectable AnalytesGentamicin, Tobramycin
Sample TypePlasma, Serum
TypeCalibrator
QuantityLow: 2 x 7.5 mL; High: 2 x 5.0 mL
Unit SizeEach
Catalog Number
100001TS
also known as 100001
Unit Size
Each
Description
CEDIA Cardiac TDM Multi-Calibrators
Detectable Analytes
NAPA
Price (USD)
Full specifications
DescriptionCEDIA Cardiac TDM Multi-Calibrators
Detectable AnalytesNAPA, Procainamide
Sample TypePlasma, Serum
TypeCalibrator
QuantityLow: 2 x 7.5 mL; High: 2 x 5.0 mL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
CEDIA Antiepileptic TDM Multi-Calibrators
Detectable Analytes
Carbamazepine
Price (USD)
Full specifications
DescriptionCEDIA Antiepileptic TDM Multi-Calibrators
Detectable AnalytesCarbamazepine, Phenobarbital, Phenytoin, Theophylline, Valproic Acid
Sample TypePlasma, Serum
TypeCalibrator
QuantityLow: 2 x 7.5 mL; High: 2 x 5.0 mL
Unit SizeEach
Showing 3 of 3
Catalog NumberSpecificationsUnit SizeDescriptionDetectable AnalytesPrice (USD)
100017Full specifications
EachCEDIA Antibiotic TDM Multi-CalibratorsGentamicinRequest A Quote
DescriptionCEDIA Antibiotic TDM Multi-Calibrators
Detectable AnalytesGentamicin, Tobramycin
Sample TypePlasma, Serum
TypeCalibrator
QuantityLow: 2 x 7.5 mL; High: 2 x 5.0 mL
Unit SizeEach
100001TS
also known as 100001
Full specifications
EachCEDIA Cardiac TDM Multi-CalibratorsNAPARequest A Quote
DescriptionCEDIA Cardiac TDM Multi-Calibrators
Detectable AnalytesNAPA, Procainamide
Sample TypePlasma, Serum
TypeCalibrator
QuantityLow: 2 x 7.5 mL; High: 2 x 5.0 mL
Unit SizeEach
100007Full specifications
EachCEDIA Antiepileptic TDM Multi-CalibratorsCarbamazepineRequest A Quote
DescriptionCEDIA Antiepileptic TDM Multi-Calibrators
Detectable AnalytesCarbamazepine, Phenobarbital, Phenytoin, Theophylline, Valproic Acid
Sample TypePlasma, Serum
TypeCalibrator
QuantityLow: 2 x 7.5 mL; High: 2 x 5.0 mL
Unit SizeEach
Showing 3 of 3
  • Proven technology
  • Liquid ready-to-use: no reconstitution needed
  • Multiple drug constituents per calibration kit
  • Positive linear calibration curve: the higher the absorbance reading, the more drug present
  • Excellent correlation to reference methods
  • Recombinant DNA technology provides lot-to-lot consistency
For In Vitro Diagnostic Use.  

Contents & Storage

2°C to 8°C