Description | Tobramycin II Assay |
Detectable Analytes | Tobramycin |
Sample Type | Plasma, Serum |
Type | Reagent |
Quantity | 13 x 11 mL |
Unit Size | Each |
Description | Carbamazepine Assay |
Detectable Analytes | Carbamazepine |
Sample Type | Plasma, Serum |
Type | Reagent |
Quantity | 17 x 17 mL |
Unit Size | Each |
Description | Gentamicin II Assay |
Detectable Analytes | Gentamicin II |
Type | Reagent |
Quantity | 13 x 11 mL |
Unit Size | Each |
Description | NAPA Assay |
Detectable Analytes | NAPA |
Sample Type | Plasma, Serum |
Type | Reagent |
Quantity | 17 x 17 mL |
Unit Size | Each |
Description | Phenobarbital II Assay |
Detectable Analytes | Phenobarbital |
Sample Type | Plasma, Serum |
Type | Reagent |
Quantity | 17 x 17 mL |
Unit Size | Each |
Description | Phenytoin II Assay |
Detectable Analytes | Phenytoin |
Sample Type | Plasma, Serum |
Type | Reagent |
Quantity | 17 x 17 mL |
Unit Size | Each |
Description | Procainamide Assay |
Detectable Analytes | Procainamide |
Sample Type | Plasma, Serum |
Type | Reagent |
Quantity | 17 x 17 mL |
Unit Size | Each |
Description | Theophylline II Assay |
Detectable Analytes | Theophylline |
Sample Type | Plasma, Serum |
Type | Reagent |
Quantity | 17 x 17 mL |
Unit Size | Each |
Description | Valproic Acid Assay |
Detectable Analytes | Valproic Acid |
Sample Type | Plasma, Serum |
Type | Reagent |
Quantity | 13 x 11 mL |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
100018 | Each | Tobramycin II Assay | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
100006 | Each | Carbamazepine Assay | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
100016 | Each | Gentamicin II Assay | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
100015 | Each | NAPA Assay | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
100003TS also known as 100003 | Each | Phenobarbital II Assay | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
100002TS also known as 100002 | Each | Phenytoin II Assay | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
100014 | Each | Procainamide Assay | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
100008 | Each | Theophylline II Assay | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
100013TS also known as 100013 | Each | Valproic Acid Assay | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|