Interested in a bulk or custom version of this product? Request pricing

EGFR Recombinant Human Protein, His Tag, Active

Catalog number: 10001H08H5

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Epidermal growth factor receptor (EGFR), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant protein

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ser 645) of human EGFR (NP_005219) fused to a polyhistidine tag at the C-terminus.

Activity: Measure by its ability to bind with human EGF in a functional ELISA.

Formulation: Lyophilized in 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween-80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 25 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

Specifications

Protein Family: Growth Factors & Receptors, Kinases & Inhibitors
Research Category: Cardiovascular Research, Oncology, Signal Transduction, Stem Cell Research
Endotoxin Level: < 1 EU/µg
Form: Lyophilized
Gene Aliases: EGFR, ErbB, HER1, mENA, ErbB1, PIG61
Gene Symbol: EGFR
Kinase Group: Tyrosine Kinases
Molecular Weight: 69.8 kDa
Product Size: 5 µg
Protein Name: Epidermal growth factor receptor
Protein Subtype: Growth Factor Receptors, Receptor Tyrosine Kinases
Protein Tag or Fusion: His Tag
Purity: > 95% by SDS-PAGE
Species: Human
Validated Application: Assay Standard
Product Line: Sino Biological®
Expression: Mammalian
Kinase Family: EGF Receptor (ErbB) Kinase Family
Protein Form: Cell Surface Protein, Recombinant, Truncated

Contents & storage

Includes 1 vial of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols

Recommended products