Attune® NxT Waste Bottle

Catalog number: 100022156

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This is a 1.9 L replacement bottle for the waste tank on the Attune® NxT Acoustic Focusing Cytometer.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Attune® Acoustic Focusing Cytometer
Product Line: Attune®
Product Size: 1 bottle
Type: Bottle

Contents & storage

Store at room temperature.

Documents

Manuals & protocols

Recommended products