Product Image
Attune™ NxT Bandpass Blade
Have Questions?
Catalog number 100022652
Price (USD)
-
This is a blank emission filter holder used to create a custom emission filter for use on the Attune® NxT Acoustic Focusing Cytometer.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Unit Size1 filter
Contents & Storage
Store at room temperature.