Product Image
Invitrogen™

Attune™ NxT Emission Filter, 603/48BP

This is a replacement 603/48 bandpass filter used in the optics path of the Attune™ NxT Acoustic Focusing Cytometer.Leia mais
Have Questions?
Número do catálogoQuantity
1000227611 filter
Número do catálogo 100022761
Preço (BRL)
-
Quantity:
1 filter
This is a replacement 603/48 bandpass filter used in the optics path of the Attune™ NxT Acoustic Focusing Cytometer.
For Research Use Only. Not for use in diagnostic procedures.
Especificações
Product LineAttune
Quantity1 filter
For Use With (Equipment)Attune Acoustic Focusing Cytometer
Product TypeEmission Filter
Unit SizeEach
Conteúdo e armazenamento
Store at room temperature.