You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Home
▸
Shop All Products
▸
Cell Analysis Products
▸
Flow Cytometry Products
▸
Flow Cytometry Analyzer Accessories
▸
Attune™ NxT Dichroic Filter, 600DLP
Invitrogen™
Attune™ NxT Dichroic Filter, 600DLP
This is a replacement 600 DLP dichroic filter used in the optics path of the Attune™ NxT Acoustic Focusing
Cytometer.
Read more
Have Questions?
Catalog Number
Quantity
100022771
1 filter
Catalog number
100022771
Price (CLP)
-
Quantity:
1 filter
This is a replacement 600 DLP dichroic filter used in the optics path of the Attune™ NxT Acoustic Focusing Cytometer.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product Line
Attune
Quantity
1 filter
For Use With (Equipment)
Attune Acoustic Focusing Cytometer
Product Type
Dichroic Filter
Unit Size
Each
Contents & Storage
Store at room temperature.