Product Image
Invitrogen™

Attune™ NxT Dichroic Filter, 690DLP

Have Questions?
Catalog NumberQuantity
1000227731 filter
Catalog number 100022773
Price (MXN)
-
Quantity:
1 filter
This is a replacement 690 DLP dichroic filter used in the optics path of the Attune® NxT Acoustic Focusing Cytometer.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineAttune
Quantity1 filter
For Use With (Equipment)Attune Acoustic Focusing Cytometer
Product TypeDichroic Filter
Unit SizeEach
Contents & Storage
Store at room temperature.