Attune™ NxT and CytPix Fluorescent Protein Filter Kit
The Attune™ NxT Fluorescent Protein Filter Kit enables the configuration of the Attune™ NxT Acoustic Focusing Cytometer for multiplex detection深入閱讀
Have Questions?
產品號碼
Quantity
100022775
1 kit
產品號碼 100022775
價格 (TWD)
99,561.00
Each
Quantity:
1 kit
價格 (TWD)
99,561.00
Each
The Attune™ NxT Fluorescent Protein Filter Kit enables the configuration of the Attune™ NxT Acoustic Focusing Cytometer for multiplex detection of GFP, YFP, and mCherry fluorescent proteins.
Optimized for fluorescent protein detection At times it can be difficult to detect multiple fluorescent proteins and maintain the best signal detection due to emission signal overlap. This filter kit has been optimized for the detection of GFP in the BL1 channel with a 510/10 BP filter off of the blue laser, YFP in the BL2 channel with a 540/30 BP filter off of the blue laser, and mCherry in the YL2 channel with a 615/25 BP filter off of the yellow laser.
For Research Use Only. Not for use in diagnostic procedures.
規格
Product LineAttune
Quantity1 kit
For Use With (Equipment)Attune Acoustic Focusing Cytometer
Product TypeProtein Filter Kit
Unit SizeEach
內容物與存放
Contains: • 1 Dichroic 525 LP filter • 1 Emission 510/10 BP filter • 1 Emission 540/30 BP filter • 1 Emission 615/25 BP filter