Attune™ Laser Upgrade Kits
Invitrogen™

Attune™ Laser Upgrade Kits

Enhance the capabilities of your Attune NxT Acoustic Focusing Flow Cytometer and Attune CytPix Imaging Enhanced Flow Cytometer with our versatile laser upgrade kits.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorFor Use With (Equipment)
100022778RedAttune Acoustic Focusing Cytometer
100022777VioletAttune Acoustic Focusing Cytometer
100022779YellowAttune Acoustic Focusing Cytometer
A32701GreenAttune Acoustic Focusing Cytometer
A52444RedAttune CytPix Flow Cytometer
A52445YellowAttune CytPix Flow Cytometer
A52446VioletAttune CytPix Flow Cytometer
Catalog number 100022778
Price (USD)
-
Color:
Red
For Use With (Equipment):
Attune Acoustic Focusing Cytometer

Enhance the capabilities of your Attune NxT Acoustic Focusing Flow Cytometer and Attune CytPix Imaging Enhanced Flow Cytometer with our versatile laser upgrade kits. Available in red, yellow, violet, and green options, these upgrade kits are designed to expand your instrument's fluorescence detection capabilities, enabling a broader range of research applications and enhancing the versatility of your flow cytometry experiments.

Expanded capabilities

Maximize your research potential with our laser upgrade kits. Each kit is tailored to specific excitation wavelengths, allowing you to utilize a variety of labels, dyes, and applications. For example, the Red Laser Upgrade Kit expands the 637-nm excitation capabilities, facilitating the detection of APC, Alexa Fluor 700, APC-tandem dyes, and functional reagents such as CellTrace Far Red dye, LIVE/DEAD Fixable far red or near IR dyes, Vybrant DyeCycle Ruby dye, and far red–exciting fluorescent proteins. Similar enhancements are available with the yellow, violet, and green laser upgrade kits, each broadening the scope of detectable fluorophores and applications.

Cost-effective solutions

Upgrading your existing flow cytometer is a cost-effective way to extend its capabilities without the need to invest in new instruments. These upgrades allow you to configure your Attune NxT and Attune CytPix flow cytometers with different laser configurations, meeting the evolving demands of your research while optimizing your budget.

Simple installation

The Attune NxT and Attune CytPix flow cytometers are designed for easy upgrades. Each laser upgrade kit includes all necessary optical components and can be installed by a qualified service engineer in typically less than four hours. This minimizes downtime and ensures that your instrument is quickly back in operation with enhanced functionality.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorRed
Product LineAttune
Quantity1 Kit
For Use With (Equipment)Attune Acoustic Focusing Cytometer
Product TypeLaser Upgrade Kit
Unit SizeEach
Contents & Storage
Contains red laser with fiber optic cable and optics block.