Interested in a bulk or custom version of this product? Request pricing.

HA1 Subunit Recombinant Influenza A Virus Protein, subtype H5N1, (A/Vietnam/1203/2004), (mIgG1-Fc.His Tag)

Catalog number: 10003V06H1250

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

H5N1, recombinant influenza A virus protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.

This recombinant protein was expressed from a DNA sequence encoding the N-terminal segment (Met 1-Arg 342) of influenza A virus, subtype H5N1 (A/VietNam/1203/2004), HA1 subunit (AAW80717.1) fused to the His-tagged Fc region of mouse IgG1 at the C-terminus.

Activity: Measured by its binding ability in a functional ELISA.

Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Microbiology, Virology
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: HA1
Gene Symbol: HA
Molecular Weight: 26.1 kDa
Product Line: Sino Biological®
Product Size: 5 x 50 µg
Protein Family: Other Proteins
Protein Form: Cell Surface Protein, Recombinant, Truncated
Protein Name: Hemagglutinin
Protein Subtype: Viral Proteins
Protein Tag or Fusion: His Tag, IgG1 Fc Tag
Purity: ≥ 85% by SDS-PAGE and SEC-HPLC Analysis.
Species: Influenza
Validated Application: Assay Standard
Shipping Condition: Room Temperature

Contents & storage

Store at -20 to -70 °C

Documents

Manuals & protocols