Product Image
Thermo Scientific™

CEDIA™ Amphetamine/Ecstasy Drugs of Abuse Assays

Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of Thermo Scientific™ CEDIA™ Amphetamine/Ecstasy Drugs of Abuse Assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
100040TS1 x 495 mL, LC
100103TS1 x 65 mL, MCC
100164173 x 17 mL, Indiko
100104TS3 x 17 mL, TSC
Catalog number 100040TS
Price (USD)
-
Quantity:
1 x 495 mL, LC

Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of Thermo Scientific™ CEDIA™ Amphetamine/Ecstasy Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA™ assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs.

  • One of the largest menus of tests available
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Control SetsMGC Multi-Drug Controls
DescriptionCEDIA Amphetamine/Ecstasy Assays
Quantity1 x 495 mL, LC
Sample TypeUrine
TypeReagent
Detectable AnalytesAmphetamine (AMP), Methylenedioxymethamphetamine (MDMA)
DoA CalibratorsMulti-drug Calibrator
Unit SizeEach
Contents & Storage
2°C to 8°C