MGC 240 Benchtop Analyzer
MGC 240 Benchtop Analyzer
MGC 240 Benchtop Analyzer
Thermo Scientific™

MGC 240 Benchtop Analyzer

Run 240 tests per hour, handling up to 24 two-reagent immunoassays per run with the Thermo Scientific™ MGC 240 Benchtop Analyzer.
Have Questions?
Catalog number 10007260
Price (USD)
-

Run 240 tests per hour, handling up to 24 two-reagent immunoassays per run with the Thermo Scientific™ MGC 240 Benchtop Analyzer. This open, flexible system can be used for drugs of abuse screens and therapeutic drug monitoring including immunosuppressive diagnostic assays and specimen validity testing.

Features

  • Broad menu of immunoassays with multiple technology options
  • Multiple channels: up to 24 two-reagent tests
  • High throughput: 240 tests/hour
  • 24-hour reagent cooling
  • Barcoding capabilities

Functions

  • Random access, true STAT interrupt capability
  • Adapts to various primary tubes and sample cups
  • Automatic cuvette QC
  • Auto rerun, auto dilution, sample pre-dilution
  • Re-suspension mixing for latex particles
  • Auto alarms for washing and waste containers
  • Air pressure mixing: no carry over

Software

  • Microsoft™ Windows™-based technology

Cuvettes

  • Unique design
  • Easy maintenance
  • Distinctive air pressure mixing system: no mixing paddles
  • No carry over: fewer washing steps
For In Vitro Diagnostic Use.  
Specifications
Amperage15 A
Analytical MethodEnd point, rate assay
CalibrationLinear, spline, logit-log, exponential, polynominal
CuvettesUnique design, no carry-over, semi-permanent
Depth (English)25 in.
Depth (Metric)63.5 cm
DescriptionMGC 240 Benchtop Analyzer
Distilled Water Consumption3.5 L/hr.
Electrical Requirements110/115/230 V 50/60 Hz
Frequency50/60 Hz
Height (English)21 in.
Height (Metric)53.3 cm
ISE TypeNot available in USA
Measurement MethodPhotometric
Operating SystemWindows™ based technology
Quality Control DetailsWestguard multi-rule
Reaction Vessel200µL minimum to 400µL maximum
ReagentsDAT, TDM, Clinical Chemistry
Sample Dilution0.048611111111
Throughput240 Tests/hr.
Voltage110/115/230 V
Weight (English)180 lb.
Weight (Metric)82 kg
Width (English)31 in.
Width (Metric)78.7 cm
Sample TypeSerum, Urine
TypeBenchtop Analyzer
Unit SizeEach