Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Type | Reagent Bottle |
Description | Reagent Bottle; 60mL; With Cap |
Capacity (Metric) | 60 mL |
Quantity | 20/Pk. |
Unit Size | Each |
Type | Reaction Cuvette |
Description | Reaction Cuvettes |
Quantity | 60/Pk. |
Unit Size | Each |
Type | Reagent Bottle |
Description | Reagent Bottle; 40mL; With Cap |
Capacity (Metric) | 40 mL |
Quantity | 20/Pk. |
Unit Size | Each |
Type | Reagent Bottle |
Description | Reagent Bottle; 20mL; With Cap |
Capacity (Metric) | 20 mL |
Quantity | 20/Pk. |
Unit Size | Each |
Type | Acid Washing Solution |
Description | Acid Washing Solution |
Detects | Cleaning Solution |
Quantity | 2 x 500 mL |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
10007625 | Each | Reagent Bottle; 60mL; With Cap | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
10007624 | Each | Reaction Cuvettes | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
10007626 | Each | Reagent Bottle; 40mL; With Cap | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
10007627 | Each | Reagent Bottle; 20mL; With Cap | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
100262TS also known as 100262 | Each | Acid Washing Solution | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|