Product Image
Thermo Scientific™

DRI™ Ecstasy Drugs of Abuse Calibrators

Utilize the globally valued accuracy, precision and lot-to-lot dependability of Thermo Scientific™ DRI™ Ecstasy Drugs of Abuse Calibrators.
Have Questions?
變更視圖buttonViewtableView
產品號碼Concentration
100080750 ng/mL
1000791000 ng/mL
100081500 ng/mL
100082250 ng/mL
產品號碼 100080
價格 (HKD)
-
Concentration:
750 ng/mL

Utilize the globally valued accuracy, precision and lot-to-lot dependability of Thermo Scientific™ DRI™ Ecstasy Drugs of Abuse Calibrators, which are optimized for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and DRI assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing on the market.

  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
規格
Concentration750 ng/mL
DescriptionDRI Ecstasy Drugs of Abuse Calibrators
Packaging1 Bottle
Quantity1 x 10 mL
TypeCalibrator
Detectable AnalytesMDA, MDEA, MDMA
Sample TypeUrine
Unit SizeEach
內容物與存放
2°C to 8°C