Product Image
Thermo Scientific™

CEDIA™ Opiate Drugs of Abuse Assays

Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of CEDIA™ Opiate Drugs of Abuse Assays.
Have Questions?
Catalog NumberQuantity
1000893 x 17 mL, TSC
Catalog number 100089
Price (MXN)
-
Quantity:
3 x 17 mL, TSC
Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of CEDIA™ Opiate Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Control SetsMGC Multi-Drug Controls
DescriptionCEDIA Opiate Drugs of Abuse Assays
Quantity3 x 17 mL, TSC
Sample TypeUrine
TypeReagent
Detectable AnalytesOpiates (OPI)
DoA CalibratorsMulti-drug Calibrator
Unit SizeEach
Contents & Storage
2°C to 8°C