Thermo Scientific™

MGC 240 Benchtop Analyzer Consumables

Catalog number: 10010235
Thermo Scientific™

MGC 240 Benchtop Analyzer Consumables

Catalog number: 10010235
Ensure uninterrupted performance of the Thermo Scientific™ MGC 240 Benchtop Analyzer with a good supply of Thermo Scientific MGC 240 Benchtop Analyzer Consumables.
 
Catalog Number
10010235
Unit Size
Each
Description
Sample Tubes; Plastic; 15 x 75mm
Price (USD)
Full specifications
TypeSample Tube
DescriptionSample Tubes; Plastic; 15 x 75mm
Quantity1/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Reaction Cuvettes
Price (USD)
Full specifications
TypeReaction Cuvette
DescriptionReaction Cuvettes
Quantity60/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Reagent Bottle; 60mL; With Cap
Price (USD)
Full specifications
TypeReagent Bottle
DescriptionReagent Bottle; 60mL; With Cap
Capacity (Metric)60 mL
Quantity20/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Reagent Bottle; 40mL; With Cap
Price (USD)
Full specifications
TypeReagent Bottle
DescriptionReagent Bottle; 40mL; With Cap
Capacity (Metric)40 mL
Quantity20/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Reagent Bottle; 20mL; With Cap
Price (USD)
Full specifications
TypeReagent Bottle
DescriptionReagent Bottle; 20mL; With Cap
Capacity (Metric)20 mL
Quantity20/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Thermal Printer Paper; 2-3/8 x 2 in.
Price (USD)
Full specifications
TypeThermal Printer Paper
DescriptionThermal Printer Paper; 2-3/8 x 2 in.
Quantity24 Rolls/Cs.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Sample Cups; Hitachi Type
Price (USD)
Full specifications
TypeHitachi Type Sample Cup
DescriptionSample Cups; Hitachi Type
QuantityPk. of 1000
Unit SizeEach
Catalog Number
100262TS
also known as 100262
Unit Size
Each
Description
Acid Washing Solution
Price (USD)
Full specifications
TypeAcid Washing Solution
DescriptionAcid Washing Solution
DetectsCleaning Solution
Quantity2 x 500 mL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Base Washing solution
Price (USD)
Full specifications
TypeBase Washing Solution
DescriptionBase Washing solution
DetectsCleaning Solution
Quantity2 x 500 mL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Sample Cup Holder
Price (USD)
Full specifications
TypeSample Cup Holder
DescriptionSample Cup Holder
For Use With (Equipment)MGC 240 Benchtop Analyzer
Quantity10/Pk.
Unit SizeEach
Showing 10 of 10
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
10010235Full specifications
EachSample Tubes; Plastic; 15 x 75mmRequest A Quote
TypeSample Tube
DescriptionSample Tubes; Plastic; 15 x 75mm
Quantity1/Pk.
Unit SizeEach
10007624Full specifications
EachReaction CuvettesRequest A Quote
TypeReaction Cuvette
DescriptionReaction Cuvettes
Quantity60/Pk.
Unit SizeEach
10007625Full specifications
EachReagent Bottle; 60mL; With CapRequest A Quote
TypeReagent Bottle
DescriptionReagent Bottle; 60mL; With Cap
Capacity (Metric)60 mL
Quantity20/Pk.
Unit SizeEach
10007626Full specifications
EachReagent Bottle; 40mL; With CapRequest A Quote
TypeReagent Bottle
DescriptionReagent Bottle; 40mL; With Cap
Capacity (Metric)40 mL
Quantity20/Pk.
Unit SizeEach
10007627Full specifications
EachReagent Bottle; 20mL; With CapRequest A Quote
TypeReagent Bottle
DescriptionReagent Bottle; 20mL; With Cap
Capacity (Metric)20 mL
Quantity20/Pk.
Unit SizeEach
10009118Full specifications
EachThermal Printer Paper; 2-3/8 x 2 in.Request A Quote
TypeThermal Printer Paper
DescriptionThermal Printer Paper; 2-3/8 x 2 in.
Quantity24 Rolls/Cs.
Unit SizeEach
10010582Full specifications
EachSample Cups; Hitachi TypeRequest A Quote
TypeHitachi Type Sample Cup
DescriptionSample Cups; Hitachi Type
QuantityPk. of 1000
Unit SizeEach
100262TS
also known as 100262
Full specifications
EachAcid Washing SolutionRequest A Quote
TypeAcid Washing Solution
DescriptionAcid Washing Solution
DetectsCleaning Solution
Quantity2 x 500 mL
Unit SizeEach
100263Full specifications
EachBase Washing solutionRequest A Quote
TypeBase Washing Solution
DescriptionBase Washing solution
DetectsCleaning Solution
Quantity2 x 500 mL
Unit SizeEach
20-30-0639Full specifications
EachSample Cup HolderRequest A Quote
TypeSample Cup Holder
DescriptionSample Cup Holder
For Use With (Equipment)MGC 240 Benchtop Analyzer
Quantity10/Pk.
Unit SizeEach
Showing 10 of 10
Ensure uninterrupted performance of the Thermo Scientific™ MGC 240 Benchtop Analyzer with a good supply of Thermo Scientific MGC 240 Benchtop Analyzer Consumables.
For In Vitro Diagnostic Use.  

Figures

Frequently asked questions (FAQs)

Citations & References