Attune™ Xenith™ 781 nm Laser Line Filters
Invitrogen™

Attune™ Xenith™ 781 nm Laser Line Filters

These replacement optical filters are for those channels available off the 781 nm laser of the Attune Xenith Flow Cytometer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberExcitation/Emission
100117900CS828DLP
100117849CS810/25
100117850CS844/25
100117851CS860/LP
100117899CS860DLP
Catalog number 100117900CS
Price (USD)
162.00
Each
Excitation/Emission:
828DLP
Price (USD)
162.00
Each
The Attune Xenith Flow Cytometer is designed with a flexible optical deck and the ability to run both spectral unmixing and conventional compensation assays on one system. Part of this flexibility is delivered through user-changeable optical filters. These optical filters are for those channels available off the 781 nm laser.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorIR
Excitation/Emission828DLP
Quantity1 Ea.
Wavelength781 nm
For Use With (Equipment)Attune Xenith Flow Cytometer
Product TypeEmission Filter
Unit SizeEach