QMS™ Tacrolimus Immunoassays and Calibrators
Actual product may vary
Thermo Scientific™
QMS™ Tacrolimus Immunoassays and Calibrators
Achieve excellent low-end precision and sensitivity with the Thermo Scientific™ QMS™ Tacrolimus Immunoassay.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
10015556Tacrolimus Immunoassay
10015573Tacrolimus Calibrators
Catalog number 10015556
Price (USD)
-
Type:
Tacrolimus Immunoassay
Achieve high sensitivity and excellent low-end precision when monitoring kidney, liver and heart transplant patients with the Thermo Scientific™ QMS™ Tacrolimus Immunoassay. With a reportable range down to 1ng/mL, our Tacrolimus Immunoassay is the perfect aid for monitoring patients on multiple drug therapies.The reagents are liquid and ready-to-use with applications available on a wide variety of other laboratory instrumentation. List of applications available upon request.
Key Features: 
  • Optimal low-end sensitivity with a broad dynamic range
  • Excellent correlation with HPLC-MS, the gold standard method
  • Precise throughout the standard curve assay range
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionQMS Tacrolimus Immunoassay for Indiko
Quantity18 mL R1, 12 mL R2, 50 mL extraction
TypeTacrolimus Immunoassay
Detectable AnalytesTacrolimus
Sample TypeWhole Blood
Unit SizeEach