QMS™ Tacrolimus Immunoassays and Calibrators
Actual product may vary
Thermo Scientific™
QMS™ Tacrolimus Immunoassays and Calibrators
Achieve excellent low-end precision and sensitivity with the Thermo Scientific™ QMS™ Tacrolimus Immunoassay.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
10015573Tacrolimus Calibrators
10015556Tacrolimus Immunoassay
Catalog number 10015573
Price (MXN)
-
Type:
Tacrolimus Calibrators
Achieve high sensitivity and excellent low-end precision when monitoring kidney, liver and heart transplant patients with the Thermo Scientific™ QMS™ Tacrolimus Immunoassay. With a reportable range down to 1ng/mL, our Tacrolimus Immunoassay is the perfect aid for monitoring patients on multiple drug therapies.The reagents are liquid and ready-to-use with applications available on a wide variety of other laboratory instrumentation. List of applications available upon request.
Key Features: 
  • Optimal low-end sensitivity with a broad dynamic range
  • Excellent correlation with HPLC-MS, the gold standard method
  • Precise throughout the standard curve assay range
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Quantity6 calibrator set: 4 mL Cal A, 2 mL Cal B to E
TypeTacrolimus Calibrators
Detectable AnalytesTacrolimus
Sample TypeWhole Blood
Unit SizeEach