DRI™ Ethyl Glucuronide (EtG) Drugs of Abuse Control Sets
DRI™ Ethyl Glucuronide (EtG) Drugs of Abuse Control Sets
Actual product may vary
Thermo Scientific™

DRI™ Ethyl Glucuronide (EtG) Drugs of Abuse Control Sets

Save time while maintaining accuracy with these liquid, ready-to-use control sets, which are for Criminal Justice and Forensic Use Only and are for sale in the U.S.A. only.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConcentration
10015936625 ng/mL
10015934375 ng/mL
10015937750 ng/mL
100159391250 ng/mL
Catalog number 10015936
Price (CLP)
-
Concentration:
625 ng/mL
Streamline your testing process while maintaining high analytical accuracy by utilizing Thermo Scientific™ DRI™ Ethyl Glucuronide (EtG) Drugs of Abuse Control Sets along with our Thermo Scientific DRI Ethyl Glucuronide (EtG) Drugs of Abuse Assays and Calibrators. These control sets are for Criminal Justice and Forensic Use Only and are for sale in the U.S.A. only.
  • Specific detection of Ethyl Glucuronide (EtG)
  • Qualitative or semi-quantitative modes
  • Excellent correlation to LC-MS/MS
  • Applications for an array of clinical chemistry analyzers
For sale in the U.S.A. only.
For Criminal Justice and Forensic Use Only.
Specifications
Concentration625 ng/mL
DescriptionDRI Ethyl Glucuronide (EtG) 625 Control. For Criminal Justice and Forensic Use Only. For sale in the U.S.A. only.
Quantity1 x 25 mL
TypeControl
Detectable AnalytesEthyl Glucuronide (EtG)
Sample TypeUrine
Unit SizeEach
Contents & Storage
2°C to 8°C