Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | DRI Fentanyl Control Set. Criminal Justice and Forensic use only. For sale in the U.S.A. only. |
Concentration | 1 ng/mL |
Detectable Analytes | Fentanyl |
Sample Type | Urine |
Type | Control |
Packaging | 1 Bottle |
Quantity | 1 x 10 mL |
Unit Size | Each |
Description | DRI Fentanyl Control Set. Criminal Justice and Forensic use only. For sale in the U.S.A. only. |
Concentration | 3 ng/mL |
Detectable Analytes | Fentanyl |
Sample Type | Urine |
Type | Control |
Packaging | 1 Bottle |
Quantity | 1 x 10 mL |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Concentration | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
10016022 | Each | 1 ng/mL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
10016024 | Each | 3 ng/mL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|