Thermo Scientific™

CEDIA™ Cocaine Drugs of Abuse Assays

Catalog number: 10016413
Thermo Scientific™

CEDIA™ Cocaine Drugs of Abuse Assays

Catalog number: 10016413
Utilize the globally-valued accuracy, precision, lot-to-lot dependability and stable shelf life of CEDIA™ Cocaine Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs.
 
Catalog Number
10016413
Unit Size
Each
Quantity
3 x 17 mL, Indiko
Price (USD)
Full specifications
Control SetsMGC Multi-Drug Controls
Quantity3 x 17 mL, Indiko
DescriptionCEDIA Cocaine Drugs of Abuse Assay for Indiko
Detectable AnalytesBenzoylecgonine
Sample TypeUrine
DoA CalibratorsCEDIA Multi-Drug Calibrators
TypeReagent
Unit SizeEach
Catalog Number
Unit Size
-
Quantity
3 x 17 mL, TSC
Price (USD)
Full specifications
Control SetsMGC Multi-Drug Controls
Quantity3 x 17 mL, TSC
DescriptionCEDIA Cocaine Drugs of Abuse Assays
Detectable AnalytesBenzoylecgonine
Sample TypeUrine
DoA CalibratorsCEDIA Multi-Drug Calibrators
TypeReagent
Unit Size-
Catalog Number
Unit Size
Each
Quantity
1 x 65 mL, MCC
Price (USD)
Full specifications
Control SetsMGC Multi-Drug Controls
Quantity1 x 65 mL, MCC
DescriptionCEDIA Cocaine Drugs of Abuse Assays
Detectable AnalytesBenzoylecgonine
Sample TypeUrine
DoA CalibratorsCEDIA Multi-Drug Calibrators
TypeReagent
Unit SizeEach
Catalog Number
Unit Size
Each
Quantity
1 x 495 mL, LC
Price (USD)
Full specifications
Control SetsMGC Multi-Drug Controls
Quantity1 x 495 mL, LC
DescriptionCEDIA Cocaine Drugs of Abuse Assays
Detectable AnalytesBenzoylecgonine
Sample TypeUrine
DoA CalibratorsCEDIA Multi-Drug Calibrators
TypeReagent
Unit SizeEach
Showing 4 of 4
Catalog NumberSpecificationsUnit SizeQuantityPrice (USD)
10016413Full specifications
Each3 x 17 mL, IndikoRequest A Quote
Control SetsMGC Multi-Drug Controls
Quantity3 x 17 mL, Indiko
DescriptionCEDIA Cocaine Drugs of Abuse Assay for Indiko
Detectable AnalytesBenzoylecgonine
Sample TypeUrine
DoA CalibratorsCEDIA Multi-Drug Calibrators
TypeReagent
Unit SizeEach
100086Full specifications
-3 x 17 mL, TSCRequest A Quote
Control SetsMGC Multi-Drug Controls
Quantity3 x 17 mL, TSC
DescriptionCEDIA Cocaine Drugs of Abuse Assays
Detectable AnalytesBenzoylecgonine
Sample TypeUrine
DoA CalibratorsCEDIA Multi-Drug Calibrators
TypeReagent
Unit Size-
100095TSFull specifications
Each1 x 65 mL, MCCRequest A Quote
Control SetsMGC Multi-Drug Controls
Quantity1 x 65 mL, MCC
DescriptionCEDIA Cocaine Drugs of Abuse Assays
Detectable AnalytesBenzoylecgonine
Sample TypeUrine
DoA CalibratorsCEDIA Multi-Drug Calibrators
TypeReagent
Unit SizeEach
1661230Full specifications
Each1 x 495 mL, LCRequest A Quote
Control SetsMGC Multi-Drug Controls
Quantity1 x 495 mL, LC
DescriptionCEDIA Cocaine Drugs of Abuse Assays
Detectable AnalytesBenzoylecgonine
Sample TypeUrine
DoA CalibratorsCEDIA Multi-Drug Calibrators
TypeReagent
Unit SizeEach
Showing 4 of 4
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For In Vitro Diagnostic Use.  

Contents & Storage

2°C to 8°C

Figures

Frequently asked questions (FAQs)

Citations & References