Thermo Scientific™

CEDIA™ THC (Cannabinoid) Drugs of Abuse Assays

Catalog number: 10016433
Have Questions?
Thermo Scientific™

CEDIA™ THC (Cannabinoid) Drugs of Abuse Assays

Catalog number: 10016433
Have Questions?
Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of Thermo Scientific™ CEDIA™ THC (Cannabinoid) Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs.
 
Catalog Number
10016433
Unit Size
Each
Quantity
3 x 17 mL, Indiko
Price (USD)
Full specifications
Control SetsCEDIA THC Controls
DescriptionCEDIA THC (Cannabinoid) Drugs of Abuse Assay for Indiko
Detectable Analytes11-nor-Δ9THC-COOH
DoA CalibratorsCEDIA Multi-level THC Calibrators
Sample TypeUrine
TypeReagent
Quantity3 x 17 mL, Indiko
Unit SizeEach
Catalog Number
Unit Size
Each
Quantity
3 x 17 mL, TSC
Price (USD)
Full specifications
Control SetsCEDIA THC Controls
DescriptionCEDIA THC (Cannabinoid) Drugs of Abuse Assays
Detectable Analytes11-nor-Δ9THC-COOH
DoA CalibratorsCEDIA Multi-level THC Calibrators
Sample TypeUrine
TypeReagent
Quantity3 x 17 mL, TSC
Unit SizeEach
Catalog Number
100100TS
also known as 100100
Unit Size
Each
Quantity
1 x 65 mL, MCC
Price (USD)
Full specifications
Control SetsCEDIA THC Controls
DescriptionCEDIA THC (Cannabinoid) Drugs of Abuse Assays
Detectable Analytes11-nor-Δ9THC-COOH
DoA CalibratorsCEDIA Multi-level THC Calibrators
Sample TypeUrine
TypeReagent
Quantity1 x 65 mL, MCC
Unit SizeEach
Catalog Number
Unit Size
Each
Quantity
1 x 495 mL, LC
Price (USD)
Full specifications
Control SetsCEDIA THC Controls
DescriptionCEDIA THC (Cannabinoid) Drugs of Abuse Assays
Detectable Analytes11-nor-Δ9THC-COOH
DoA CalibratorsCEDIA Multi-level THC Calibrators
Sample TypeUrine
TypeReagent
Quantity1 x 495 mL, LC
Unit SizeEach
Showing 4 of 4
Catalog NumberSpecificationsUnit SizeQuantityPrice (USD)
10016433Full specifications
Each3 x 17 mL, IndikoRequest A Quote
Control SetsCEDIA THC Controls
DescriptionCEDIA THC (Cannabinoid) Drugs of Abuse Assay for Indiko
Detectable Analytes11-nor-Δ9THC-COOH
DoA CalibratorsCEDIA Multi-level THC Calibrators
Sample TypeUrine
TypeReagent
Quantity3 x 17 mL, Indiko
Unit SizeEach
100091Full specifications
Each3 x 17 mL, TSCRequest A Quote
Control SetsCEDIA THC Controls
DescriptionCEDIA THC (Cannabinoid) Drugs of Abuse Assays
Detectable Analytes11-nor-Δ9THC-COOH
DoA CalibratorsCEDIA Multi-level THC Calibrators
Sample TypeUrine
TypeReagent
Quantity3 x 17 mL, TSC
Unit SizeEach
100100TS
also known as 100100
Full specifications
Each1 x 65 mL, MCCRequest A Quote
Control SetsCEDIA THC Controls
DescriptionCEDIA THC (Cannabinoid) Drugs of Abuse Assays
Detectable Analytes11-nor-Δ9THC-COOH
DoA CalibratorsCEDIA Multi-level THC Calibrators
Sample TypeUrine
TypeReagent
Quantity1 x 65 mL, MCC
Unit SizeEach
1661256Full specifications
Each1 x 495 mL, LCRequest A Quote
Control SetsCEDIA THC Controls
DescriptionCEDIA THC (Cannabinoid) Drugs of Abuse Assays
Detectable Analytes11-nor-Δ9THC-COOH
DoA CalibratorsCEDIA Multi-level THC Calibrators
Sample TypeUrine
TypeReagent
Quantity1 x 495 mL, LC
Unit SizeEach
Showing 4 of 4
  • One of the largest menu of tests available
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  

Contents & Storage

2°C to 8°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text