DRI™ Fentanyl CE Drugs of Abuse Calibrator
Thermo Scientific™

DRI™ Fentanyl CE Drugs of Abuse Calibrator

Save time while maintaining accuracy with this liquid, ready-to-use calibrator, which is for International use only and is not for sale in the U.S.A.
Have Questions?
Catalog NumberQuantity
100164851 x 10 mL
Catalog number 10016485
Price (HKD)
-
Quantity:
1 x 10 mL

Ensure accuracy and precision of your drug testing results by partnering this liquid, ready-to-use Thermo Scientific™ DRI™ Fentanyl Drugs of Abuse Calibrator with our Thermo Scientific DRI Fentanyl Drugs of Abuse Assays and Control Sets. This calibrator is for International use only and is not for sale in the U.S.A.

  • Specific detection of Fentanyl
  • Qualitative or semi-quantitative modes
  • Excellent correlation to LC-MS/MS
  • Applications for an array of clinical chemistry analyzers
For International use only. Not for sale in the U.S.A.
For In Vitro Diagnostic Use.  
Specifications
Concentration2 ng/mL
DescriptionDRI Fentanyl CE Calibrator. International use only. Not for sale in the U.S.A.
Packaging1 Bottle
Quantity1 x 10 mL
TypeCalibrator
Detectable AnalytesFentanyl
Sample TypeUrine
Unit SizeEach
Contents & Storage
2°C to 8°C