DRI™ Fentanyl CE Drugs of Abuse Control Sets
Thermo Scientific™

DRI™ Fentanyl CE Drugs of Abuse Control Sets

Save time while maintaining accuracy with these liquid, ready-to-use control sets, which are for International use only and are not for sale in the U.S.A.
Have Questions?
變更視圖buttonViewtableView
產品號碼Concentration
100164863 ng/mL cutoff
100164841 ng/mL cutoff
產品號碼 10016486
價格 (HKD)
-
Concentration:
3 ng/mL cutoff
Streamline your testing process while maintaining high analytical accuracy by utilizing Thermo Scientific™ DRI™ Fentanyl Drugs of Abuse Control Sets along with our Thermo Scientific DRI Fentanyl Drugs of Abuse Assays and Calibrators. These control sets are for International use only and are not for sale in the U.S.A.
  • Specific detection of Fentanyl
  • Qualitative or semi-quantitative modes
  • Excellent correlation to LC-MS/MS
  • Applications for an array of clinical chemistry analyzers
For international use only. Not for sale in the U.S.A.
For In Vitro Diagnostic Use.  
規格
Concentration3 ng/mL cutoff
DescriptionDRI Fentanyl CE Control Set. For international use only. Not for sale in the U.S.A.
LevelLow (225ng/mL); High (375ng/mL)
Packaging1 Bottle
Quantity1 x 10 mL
TypeControl
Detectable AnalytesFentanyl
Sample TypeUrine
Unit SizeEach
內容物與存放
2°C to 8°C