Product Image
Thermo Scientific™

CEDIA™ Phencyclidine (PCP) Drugs of Abuse Assays

Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of CEDIA™ Phencyclidine (PCP) Drugs of Abuse Assays.
Have Questions?
變更視圖buttonViewtableView
產品號碼Quantity
1001723 x 17 mL, TSC
1001731 x 65 mL, MCC
18157841 x 495 mL, LC
產品號碼 100172
價格 (HKD)
-
Quantity:
3 x 17 mL, TSC
Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of CEDIA™ Phencyclidine (PCP) Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
規格
Control SetsMGC Multi-Drug Controls
DescriptionCEDIA Phencycidine (PCP) Assay
Quantity3 x 17 mL, TSC
Sample TypeUrine
TypeReagent
Detectable AnalytesPhencyclidine (PCP)
DoA CalibratorsMulti-drug Calibrator
Unit SizeEach
內容物與存放
2°C to 8°C