Product Image
Thermo Scientific™
DRI™ Cotinine Drugs of Abuse Assay
Achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation with Thermo Scientific™ DRI Cotinine Drugs of Abuse Assay.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
100185163 x 18 mL
03941 x 100 mL
03951 x 500 mL
Catalog number 10018516
Price (EUR)
-
Quantity:
3 x 18 mL
Achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation with Thermo Scientific™ DRI™ Cotinine Drugs of Abuse Assay, which offers applications for an array of clinical chemistry analyzers. This liquid, ready-to-use assay has been field-tested on a wide range of analyzers. It is enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionDRI Cotinine Drugs of Abuse Assay for Indiko
Quantity3 x 18 mL
Sample TypeUrine
Shelf LifeUntil Label Expiry Date
TypeReagent
Detectable AnalytesCotinine
Unit SizeEach
Contents & Storage
2°C to 8°C