Thermo Scientific™
CEDIA™ Buprenorphine II Drugs of Abuse Calibrators and Controls
Related applications:
Clinical
Achieve qualitative and semi-quantitative test results with the Thermo Scientific™ CEDIA™ Buprenorphine II Calibrators and Controls. The five calibrators, including negative, are used to establish a standard curve that correlates the reaction signals from the analyzer with the calibrators’ respective concentrations (in ng/mL). Two controls, one above and one below the cutoff (10ng/mL), validate the calibration curve. The calibrators and controls also provide a reference point for qualitative (positive or negative results) and semi-quantitative detection.
Back to top
  • Ensures assay accuracy
  • 18 months shelf life
  • Supports qualitative and semi-quantitative measurements
For In Vitro Diagnostic Use.