CEDIA™ Buprenorphine II Drugs of Abuse Calibrators and Controls
El producto real puede variar
Thermo Scientific™

CEDIA™ Buprenorphine II Drugs of Abuse Calibrators and Controls

Monitor assay performance with the Thermo Scientific™ CEDIA™ Buprenorphine II Drugs of Abuse Calibrators and Controls.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoDescripción
10020801Calibrador CEDIA buprenorfina II 50 ng/mL
10020800Calibrador CEDIA buprenorfina II 20 ng/mL
10020799Calibrador de valor de corte CEDIA buprenorfina II 10 ng/mL
10021390Calibrador II negativo CEDIA
Número de catálogo 10020801
Precio (USD)
-
Descripción:
Calibrador CEDIA buprenorfina II 50 ng/mL
Achieve qualitative and semi-quantitative test results with the Thermo Scientific™ CEDIA™ Buprenorphine II Calibrators and Controls. The five calibrators, including negative, are used to establish a standard curve that correlates the reaction signals from the analyzer with the calibrators’ respective concentrations (in ng/mL). Two controls, one above and one below the cutoff (10ng/mL), validate the calibration curve. The calibrators and controls also provide a reference point for qualitative (positive or negative results) and semi-quantitative detection.
  • Ensures assay accuracy
  • 18 months shelf life
  • Supports qualitative and semi-quantitative measurements
Para uso diagnóstico in vitro
Para uso diagnóstico in vitro
Especificaciones
Concentración50 ng/mL
DescripciónCalibrador CEDIA buprenorfina II 50 ng/mL
Cantidad1 x 5 mL
TipoCalibrator
Analitos detectablesBuprenorphine
Tipo de muestraUrine
Unit SizeEach
Contenido y almacenamiento
2°C to 8°C