CEDIA™ Oral Fluid Drugs of Abuse Assays (CJ&F)
Actual product may vary
Thermo Scientific™
CEDIA™ Oral Fluid Drugs of Abuse Assays (CJ&F)
Accurately detect barbiturates, benzodiazepines, methadone, oxycodone, or buprenorphine in oral fluid specimens. For Criminal Justice and Forensic use only in the USA.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionDetectable Analytes
10022348CEDIA Barbiturates OFT Assay. For Criminal Justice and Forensic use only. For sale in U.S.A. only.Secobarbital
10022349CEDIA Benzodiazepines OFT Assay. For Criminal Justice and Forensic use only. For sale in U.S.A. only.Nordiazepam
10022350CEDIA Methadone OFT Assay. For Criminal Justice and Forensic use only. For sale in U.S.A. only.Methadone (MTD)
10022351CEDIA Oxycodone OFT Assay. For Criminal Justice and Forensic use only. For sale in U.S.A. only.Oxycodone (OXY)
10022352CEDIA Buprenorphine OFT Assay. For Criminal Justice and Forensic use only. For sale in U.S.A. only.Buprenorphine (BUP)
Catalog number 10022348
Price (USD)
-
Description:
CEDIA Barbiturates OFT Assay. For Criminal Justice and Forensic use only. For sale in U.S.A. only.
Detectable Analytes:
Secobarbital
Utilize the globally valued accuracy, precision, lot-to-lot dependability, and stable shelf-life of Thermo Scientific™ CEDIA™ Oral Fluid Assays, which offer applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and by developing new assays to respond to ever-emerging drug screening needs. These assays are for Criminal Justice and Forensic Use Only and are for sale in the U.S.A. only.
  • One of the largest menus of tests available
  • Qualitative option for results
  • Excellent correlation to LC-MS/MS
  • Variety of calibrators and multi-constituent control sets
For sale in the U.S.A. only.
For Criminal Justice and Forensic Use Only.
Specifications
Control SetsCEDIA Multi-Drug OFT Controls Set B
DescriptionCEDIA Barbiturates OFT Assay. For Criminal Justice and Forensic use only. For sale in U.S.A. only.
Quantity1 x 65 mL
TypeReagent
Detectable AnalytesSecobarbital
DoA CalibratorsMulti-drug OFT Cutoff Calibrator Set B
Unit SizeEach
Contents & Storage
2°C to 8°C