CEDIA™ Buprenorphine Drugs of Abuse Assays
Actual product may vary
CEDIA™ Buprenorphine Drugs of Abuse Assays
CEDIA™ Buprenorphine Drugs of Abuse Assays
CEDIA™ Buprenorphine Drugs of Abuse Assays
Thermo Scientific™
CEDIA™ Buprenorphine Drugs of Abuse Assays
Utilize the globally-valued accuracy, precision, lot-to-lot dependability and stable shelf life of CEDIA Buprenorphine Drugs of Abuse Assays
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
100240TS
also known as 100240
1 x 65 mL, MCC
100190TS3 x 17 mL, TSC
100156583 x 17 mL, Indiko
Catalog number 100240TS
also known as 100240
Price (USD)
-
Quantity:
1 x 65 mL, MCC
Utilize the globally-valued accuracy, precision, lot-to-lot dependability and stable shelf life of Thermo Scientific™ CEDIA™ Buprenorphine Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs.
  • One of the largest menus of tests available
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Control SetsCEDIA Buprenorphine Control Set
DescriptionCEDIA Buprenorphine Drugs of Abuse Assays
Quantity1 x 65 mL, MCC
Sample TypeUrine
TypeReagent
Detectable AnalytesBuprenorphine (BUP)
DoA CalibratorsBuprenorphine Calibrator
Unit SizeEach
Contents & Storage
2°C to 8°C