Thermo Scientific™

DRI™ MDA 650ng/mL Control Kit, 25mL

Catalog number: 10024435
Thermo Scientific™

DRI™ MDA 650ng/mL Control Kit, 25mL

Catalog number: 10024435
Monitor Thermo Scientific™ DRI™ Ecstasy Plus Drugs of Abuse Assay performance with the Thermo Scientific DRI MDA 650ng/mL Control Kit.
 
Catalog Number
10024435
Unit Size
Each
Price (USD)
Full specifications
DescriptionSupports new SAMHSA guidelines (effective October 1, 2017).
Detectable AnalytesMDA
Sample TypeUrine
TypeControl
Quantity1 x 25 mL
Shelf LifeUntil Label Expiry Date
StabilityUntil label expiry date
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizePrice (USD)
10024435Full specifications
EachRequest A Quote
DescriptionSupports new SAMHSA guidelines (effective October 1, 2017).
Detectable AnalytesMDA
Sample TypeUrine
TypeControl
Quantity1 x 25 mL
Shelf LifeUntil Label Expiry Date
StabilityUntil label expiry date
Unit SizeEach
Showing 1 of 1
  • Supports new NLCP guidelines for SAMHSA Labs
  • Detects for MDA cross-reactivity of ≥ 80%
For In Vitro Diagnostic Use.  

Contents & Storage

2°C to 8°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text