DRI™ Oxycodone Drugs of Abuse Control Set
DRI™ Oxycodone Drugs of Abuse Control Set
Actual product may vary
Thermo Scientific™

DRI™ Oxycodone Drugs of Abuse Control Set

Eliminate time-consuming steps while achieving high analytical accuracy when you choose the ready-to-use Thermo Scientific™ DRI™ Oxycodone Drugs of Abuse Control Set.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConcentration
100255300 ng/mL
100254100 ng/mL
Catalog number 100255
Price (EUR)
-
Request A Quote
Concentration:
300 ng/mL
Eliminate time-consuming steps while achieving high analytical accuracy when you choose the ready-to-use Thermo Scientific™ DRI™ Oxycodone Drugs of Abuse Control Set in liquid form. These assays have been field-tested on a wide range of analyzers, and they are enthusiastically accepted in laboratories where cost-control and optimal productivity are key issues.
  • One of the largest menus of tests available
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Concentration300 ng/mL
DescriptionDRI Oxycodone Drugs of Abuse Control Set
Quantity2 x 10 mL
TypeControl
Detectable AnalytesOxycodone
Sample TypeUrine
Unit SizeEach