CEDIA™ Mycophenolic Acid Immunoassays, Calibrators and Controls
Actual product may vary
CEDIA™ Mycophenolic Acid Immunoassays, Calibrators and Controls
CEDIA™ Mycophenolic Acid Immunoassays, Calibrators and Controls
Thermo Scientific™
CEDIA™ Mycophenolic Acid Immunoassays, Calibrators and Controls
Complete your ISD panel with the Thermo Scientific™ CEDIA™ Mycophenolic Acid Immunoassay.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
100277CEDIA Mycophenolic Acid Calibrator Set
100276CEDIA Mycophenolic Acid Assay
100278TS
also known as 100278
MAS Mycophenolic Acid Control 1
100279MAS Mycophenolic Acid Control 2
100280MAS Mycophenolic Acid Control 3
Catalog number 100277
Price (USD)
-
Description:
CEDIA Mycophenolic Acid Calibrator Set
Complete your immunosuppressant drug testing panel with the Thermo Scientific™ CEDIA™ Mycophenolic Acid Immunoassay. Like our other ISD assays, the CEDIA Mycophenolic Acid Immunoassay is accurate and precise, providing users with confidence in their results. Applications are available on our flexible and intuitive Indiko™ and Indiko Plus clinical and specialty chemical systems as well as a wide variety of other laboratory instrumentation.
Key Features
  • Optimal low-end sensitivity with a broad dynamic range
  • Excellent correlation with HPLC-MS, the gold standard method
  • Precise throughout the standard curve assay range
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionCEDIA Mycophenolic Acid Calibrator Set
Quantity5 mL, 2 each level
TypeCalibrator
Detectable AnalytesMycophenolic Acid
Sample TypePlasma
Unit SizeEach
Contents & Storage
2°C to 8°C