DRI Tricyclics Serum Toxicology Assays
Thermo Scientific™
DRI Tricyclics Serum Toxicology Assays
Quickly and accurately identify tricyclics in serum samples with Thermo Scientific™ DRI™ Tricyclics Serum Toxicology Assays, excellent tools for emergency rooms when urine specimens are not conveniently available.
Have Questions?
카탈로그 번호수량
100288271 X 500 mL
카탈로그 번호 10028827
제품 가격(KRW)
-
수량:
1 X 500 mL
Quickly and accurately identify tricyclics in serum samples with Thermo Scientific™ DRI™ Tricyclics Serum Toxicology Assays, excellent tools for emergency rooms when urine specimens are not conveniently available.
These Serum Toxicology Tricyclics Assays have the flexibility of analyzing serum or urine samples. These assays are provided in convenient packaging with separate calibrators for ease of use. A multi-analyte Serum Tox Control is also available. The assays are liquid, ready-to-use and have the versatility to be run on a variety of clinical chemistry analyzers.
• Serum, plasma-and urine-based assay
• Used to identify class of drug overdose
For sale in the U.S.A. and Canada only.
체외진단 전용.
Reagents are ready-to-use and no additional preparation is needed.
사양
설명DRI Tricyclics Serum Toxicology Assays
수량1 X 500 mL
샘플 종류Serum, Plasma, Urine
유형Reagent
검출 가능한 분석물질Tricyclic Antidepressants (TCA)
Unit SizeEach