Interested in a bulk or custom version of this product? Request pricing.

TNFRSF9 Recombinant Human Protein, hIgG1-Fc.His Tag

Catalog number: 10041H03H5

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Tumor Necrosis Factor Receptor Super Family 9 (TNFRSF9), recombinant human/mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the N-terminal fragment (Met 1-Gln 186) of the extracellular domain of human TNFRSF9 (NP_001552.2) fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.

Activity: Measured by its binding ability in a functional ELISA
. Immobilized recombinant mouse 4-1BB Ligand at 20µg/ml (100 µl/well) can bind human 4-1BB with a linear range of 15.6-500 ng/ml
.

Formulation: Lyophilized in 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Drug Development, Immunology, Oncology
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: 4-1BB, CD137, CDw137, ILA, MGC2172, TNFRSF9
Gene Symbol: TNFRSF9
Molecular Weight: 45.2 kDa
Product Line: Sino Biological®
Product Size: 5 µg
Protein Family: CD Proteins, Cytokines & Receptors
Protein Name: Tumor necrosis factor receptor superfamily, member 9
Protein Subtype: TNF Receptors
Protein Tag or Fusion: His Tag, IgG1 Fc Tag
Purity: ≥ 90% by SDS-PAGE
Species: Human
Validated Application: Assay Standard

Contents & storage

Includes 1 vial of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols

Recommended products