Interested in a bulk or custom version of this product? Request pricing.

BCL2L2 Recombinant Human Protein, His tag

Catalog number: 10059H08E50

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

BCL2-Like 2 (BCL2L2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the human BCL-W (Q92843-1) (Met 1-Thr 172) fused to a polyhistidine tag at the C-terminus.

Activity: The activity of this protein has not been tested.

Formulation: 25 mM Hepes, 100 mM KCl, 10% Glycerol, pH 7.4.

Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2 mg/mL or lower. It is recommended that the protein be aliquoted and used as soon as possible. Store aliquots under sterile conditions at -5 to -30°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Expression: E. coli
Form: Frozen
Gene Aliases: BCL2L2, BCL-W, BCL2-L-2, BCLW, KIAA0271
Gene ID: 599
Gene Symbol: BCL2L2
Molecular Weight: 20 kDa
Product Line: Sino Biological®
Product Size: 50 µg
Protein Family: Other Proteins
Protein Form: Full Length, Recombinant
Protein Name: BCL2-like 2
Protein Tag or Fusion: His Tag
Purification: PAGE
Purity: ≥ 90% by SDS-PAGE
Research Category: Apoptosis, Cell Cycle & Proliferation, Oncology
Species: Human
Validated Application: Assay Standard

Contents & storage

Includes 1 vials of protein in suspension buffer. Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products