Interested in a bulk or custom version of this product? Request pricing.

CDC42 Recombinant Human Protein, GST Tag

Catalog number: 10062H09E250

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Cell division cycle 42 (CDC42), GTP binding protein of 25kDa, recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the mature form (Met 1-Cys 188) of human CDC42 isoform 2 (P60953-2) fused to the GST tag at the N-terminus.

Activity: This recombinant protein has not been tested.

Formulation: Lyophilized in 20 mM Tris buffer, 150 mM NaCl, 0.5mM reduced Glutathione, pH 8.0. Contains 5% Mannitol, 5% Trehalose, and 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Expression: E. coli
Form: Lyophilized
Gene Aliases: RP1-224A6.5, CDC42Hs, G25K
Gene ID: 998
Gene Symbol: CDC42
Molecular Weight: 48.1 kDa
Product Line: Sino Biological®
Product Size: 5 x 50 µg
Protein Family: Other Proteins
Protein Form: Recombinant
Protein Name: cell division cycle 42
Protein Subtype: GTPases
Protein Tag or Fusion: GST Tag
Purity: > 85% by SDS-PAGE
Research Category: Cardiovascular Research, Cell Cycle & Proliferation, Differentiation, Neurobiology, Oncology
Species: Human
Validated Application: Assay Standard
Shipping Condition: Room Temperature

Contents & storage

Store at -20 to -70 °C

Documents

Manuals & protocols