CREB1 (CREB) Recombinant Human Protein, N-His.MBP Tag

Catalog number: 10074H10E50

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

CAMP Responsive Element Binding protein 1 (CREB1), recombinant human protein is supplied as a lyophilized powder.

This recombinant protein was expressed from a DNA sequence encoding the human CREB1 isoform B (P16220-2) (Met 1-Asp 327) fused to an N-terminal polyhistidine-tagged MBP tag at the N-terminus.

Activity: This recombinant protein has not been tested.

Formulation: 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by SINO Biological.

Specifications

Protein Family: Transcription Factors
Expression: E. coli
Form: Lyophilized
Gene Aliases: CREB, MGC9284
Gene ID: 1385
Gene Symbol: CREB1
Molecular Weight: 78.7 kDa
Product Line: Sino Biological®
Product Size: 50 µg
Protein Form: Full Length, Recombinant
Protein Name: CAMP Responsive Element Binding protein 1
Protein Tag or Fusion: His Tag, MBP (Maltose Binding Protein)
Purification: PAGE
Purity: ≥ 90% by SDS-PAGE
Species: Human
Tag Position: N-Terminal
Validated Application: Assay Standard

Contents & storage

Includes 1 vial of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols