Interested in a bulk or custom version of this product? Request pricing.

GPC5 Recombinant Human Protein, His Tag

Catalog number:  10079H08B25

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Glypican 5 (GPC5), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the human GPC5 (NP_004457.1) (Met 1-Thr 554) fused to the polyhistidine tag at the C-terminus.

Activity: Measured by its binding ability in a functional ELISA . Immobilized human GPC5 at 5 µg/ml (100 µl/well) can bind human FGF2 with a linear range of 0.156-2.5 ng/ml .

Formulation: Lyophilized in 50mM Tris, 100mM Nacl , pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween-80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 25 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by SINO Biological.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Endotoxin Level: < 1 EU/µg
Expression: Insect
Form: Frozen
Gene Aliases: GPC5, bA93M14.1, glypican proteoglycan 5, glypican-5
Gene Symbol: GPC5
Molecular Weight: 60.5 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Family: Other Proteins
Protein Name: glypican 5
Protein Subtype: Proteoglycans
Protein Tag or Fusion: His Tag
Purity: ≥ 90% by SDS-PAGE
Research Category: Development, Drug Development, Oncology, Signal Transduction
Species: Human
Validated Application: Assay Standard

Contents & storage

Store at -80 °C.

Documents

Manuals & protocols