Interested in a bulk or custom version of this product? Request pricing.

GPC3 Recombinant Human Protein, His Tag

Catalog number: 10088H08H25

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Glypican 3 (GPC3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the human GPC3 (P51654-1) (Met 1-His 559) fused to a C-terminal polyhistidine tag.

Activity: Measured by its binding ability in a functional ELISA. Immobilized human GPC3 at 5 µg/ml (100 µl/well) can bind human bFGF / FGF2 with a linear ranger of 0.156-2.5 ng/ml.

Formulation: Lyophilized in 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: Cell Structure⁄Organelle Proteins
Research Category: Biomarker-Related Pathway(s), Development, Oncology
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: GPC3, AC002420.1, DGSX, GTR2-2, MXR7, OCI-5, SDYS, SGB, SGBS, SGBS1
Gene Symbol: GPC3
Molecular Weight: 62 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Name: Glypican 3
Protein Subtype: Proteoglycans
Protein Tag or Fusion: His Tag
Purity: ≥ 85% by SDS-PAGE
Species: Human
Validated Application: Assay Standard

Contents & storage

Includes 5 vial of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols