Smart-Vue Pro Remote Monitoring Cellular Data Plans
Thermo Scientific™
Smart-Vue Pro Remote Monitoring Cellular Data Plans
Smart-Vue Pro laboratory remote monitoring system is ideal for regulated environments in need of 21 CFR Part 11 support with audit trail. The IT friendly LoRaWan system provides easy set-up for a reliable, independent and secure wireless network.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoIncluye
100D1Y4GDATANAPlan de datos móviles para un máximo de 100 dispositivos/1 año, sin tarjeta SIM, Norteamérica
10D1Y4GDATANAPlan de datos móviles para un máximo de 10 dispositivos/1 año, sin tarjeta SIM, Norteamérica
20D1Y4GDATANAPlan de datos móviles para un máximo de 20 dispositivos/1 año, sin tarjeta SIM, Norteamérica
50D1Y4GDATANAPlan de datos móviles para un máximo de 50 dispositivos/1 año, sin tarjeta SIM, Norteamérica
250D1Y4GDATANAPlan de datos móviles para un máximo de 250 dispositivos/1 año, sin tarjeta SIM, Norteamérica
500D1Y4GDATANAPlan de datos móviles para un máximo de 500 dispositivos/1 año, sin tarjeta SIM, Norteamérica
1000D1Y4GDATANAPlan de datos móviles para un máximo de 1000 dispositivos/1 año, sin tarjeta SIM, Norteamérica
1500D1Y4GDATANAPlan de datos móviles para un máximo de 1500 dispositivos/1 año, sin tarjeta SIM, Norteamérica
2000D1Y4GDATANAPlan de datos móviles para un máximo de 2000 dispositivos/1 año, sin tarjeta SIM, Norteamérica
Número de catálogo 100D1Y4GDATANA
Precio (MXN)
-
Incluye:
Plan de datos móviles para un máximo de 100 dispositivos/1 año, sin tarjeta SIM, Norteamérica
Smart-Vue Pro laboratory remote monitoring system is ideal for regulated environments in need of 21 CFR Part 11 support with audit trail. The IT friendly LoRaWan system provides easy set-up for a reliable, independent and secure wireless network.
Hardware
Receivers
Multitech Advanced receivers (blue) support up to 1000 data logging sensors. Gateways are available in 915 MHz for North America and 865-868 MHz for rest of world.
Multitech Pro receivers (white) Support for up to 50 data logging sensors per gateway and are available in Ethernet and 4G options in 915 MHz for North America and 865-868 MHz for rest of world.
modules
Smart-Vue Pro modules collect data from independent sensors connected to equipment from any OEM. Smart-Vue Pro modules are available in two configurations- Duo and Quatro. The Duo allows up to 2 sensor channels for monitoring. The Quatro has extended range of 4 sensor channels for 2 wired and 2 wireless sensors. Ideal for monitoring several pieces of equipment simultaneously or multiple points within one piece of equipment.
Sensors
The Smart-Vue Pro sensors collect data points to be logged from any laboratory equipment OEM. Sensor kits, including gateways are available.
Subscription
Requires subscription
Availability
Check with your specialist for country availability. Receivers are available in 865-868 MHz and 915 MHz.
Especificaciones
Memoria de datosNA
Para utilizar con (equipo)Smart-Vue Pro Remote Monitoring
Clasificación IPNone
IncluyePlan de datos móviles para un máximo de 100 dispositivos/1 año, sin tarjeta SIM, Norteamérica
Cantidad1
Tipos de alarmaNone
TipoSubscription Service
Unit SizeEach