Orion™ Universal Power Adapter for Star A Meters
Thermo Scientific™
Orion™ Universal Power Adapter for Star A Meters
Ensure a constant power supply to your meter with the Thermo Scientific™ Orion™ Star Series Universal Power Adapter for Orion™ Star A series and legacy Orion™ Star series meters
Have Questions?
Catalog NumberForUseWithEquipment
1010003
Catalog number 1010003
Price (MXN)
-
Ensure a constant power supply to your meter using a spare or replacement Thermo Scientific™ Orion™ Universal Power Adapter for the Thermo Scientific™ Orion™ Star A series (Star A320, Star A220, Star A210, Star A120 and Star A110 series) benchtop and portable meters.
  • Universal 110/2220V 50/60Hz adapter
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionUniversal Power Adapter
Electrical Requirements110/220 V, 50/60 Hz
Frequency50/60 Hz
IncludesUniversal power adapter with US, EU, UK and China plug plates
Voltage110/220 V
TypeMeter Accessories
Unit SizeEach