This product is prepared by covalently coupling highly purified (>98% by SDS-PAGE) Staph. aureus Cowan strain 1 Protein A toRead more
Have Questions?
Change view
Catalog Number
Quantity
101041
5 mL
101042
25 mL
101090
also known as 10-1090
1 L
3 Options
Catalog number 101041
Price (USD)
380.65
Online exclusive
409.00
Save 28.35
Each
Quantity:
5 mL
Request bulk or custom format
Price (USD)
380.65
Online exclusive
409.00
Save 28.35
Each
This product is prepared by covalently coupling highly purified (>98% by SDS-PAGE) Staph. aureus Cowan strain 1 Protein A to Sepharose™ 4B.
This product is supplied as a 50% suspension consisting of 25ml Protein A-Sepharose™ 4B and 25 ml of PBS (10 mM phosphate-buffered saline, pH 7.4, containing 0.19% Kathon CG/ICP).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormSuspension
Quantity5 mL
Column TypeAffinity
Product LineSepharose
Product TypeProtein A Sepharose
Stationary PhaseProtein A
Unit SizeEach
Contents & Storage
This product is supplied as a 50% suspension (5 ml Protein A-Sepharose™ 4B + 5 ml PBS) Wet Ice
Citations & References (1)
Citations & References
Abstract
Interaction and regulation of subcellular localization of CED-4 by CED-9.
Authors:Wu D, Wallen HD, Nuñez G
Journal:Science
PubMed ID:9027313
The Caenorhabditis elegans survival gene ced-9 regulates ced-4 activity and inhibits cell death, but the mechanism by which this occurs is unknown. Through a genetic screen for CED-4-binding proteins, CED-9 was identified as an interacting partner of CED-4. CED-9, but not loss-of-function mutants, associated specifically with CED-4 in yeast or ... More