Inquire about OEM or Commercial Supply version of this product here.
Dynabeads™ His-Tag Isolation and Pulldown
Invitrogen™

Dynabeads™ His-Tag Isolation and Pulldown

Dynabeads™ His-Tag Isolation and Pulldown beads are 1-μm monosized, monodispersed, superparamagnetic beads with an NTA-Cobalt-based ligand which has a highRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
10104D10 mL
10103D2 mL
Catalog number 10104D
Price (USD)
1,226.65
Online exclusive
1,278.00
Save 51.35
Each
Quantity:
10 mL
Price (USD)
1,226.65
Online exclusive
1,278.00
Save 51.35
Each

Dynabeads™ His-Tag Isolation and Pulldown beads are 1-μm monosized, monodispersed, superparamagnetic beads with an NTA-Cobalt-based ligand which has a high affinity towards His-tags and similar structures on molecular targets. Their small but uniform bead size presents a high surface area per mg beads and a correspondingly high capacity for the target molecule. They enable single-step isolation of tagged molecules in manual or automated workflows that helps ensure good recovery of the target. Dynabeads His-Tag Isolation and Pulldown beads can also be a great starting point for other advanced experimental approaches where solid magnetic support is needed.

Benefits of Dynabeads His-Tag and Pulldown beads include:
• Performance—high sensitivity and reproducibility securing consistent results
• Efficiency—fast binding kinetics and high binding capacity
• Flexibility—flexible protocols with gentle and efficient liquid-phase reaction kinetics
• Automation-ready—automation protocols available with the KingFisher Purification instruments

Good signal-to-noise for numerous applications
Dynabeads His-Tag Isolation and Pulldown beads are an ideal tool for isolation of tagged recombinant proteins. They are also suited for other applications with high signal-to-noise requirements. Using the principles of immobilized metal affinity binding together with the inert surface of Dynabeads magnetic beads helps ensure a clean capture with extremely low carryover of unwanted proteins. Since the bead surface is loaded with cobalt ions, it is also possible to make a stable and lasting bond with the target molecule through oxidation if the downstream applications require it. The beads are added to the solution containing the target and mixed for 5-10 minutes. After washing, the target is eluted off and ready for downstream applications.

More than just a pulldown
In addition to being a great tool for protein pulldown and isolation, the Dynabeads His-Tag Isolation and Pulldown beads can serve as a platform for target immobilization. The bead carries cobalt Co2+ ions, so it is possible to cement the bond between the bead and the target just by using mildly oxidative conditions, such as 10 mM hydrogen peroxide for 30–60 minutes. This oxidizes cobalt to the Co3+ state, making the connection between the bead and the target permanent. It will not elute in the commonly used elution buffers for IMAC. This can later be used for applications requiring a strongly bound target, without the risk of reduced target yield.

Automation-ready for use with KingFisher purification systems
The 1-μm magnetic beads are ideal for high-throughput enrichment. This process can be automated using any of our KingFisher sample purification systems or liquid handlers.

Commercial supply 
Our manufacturing sites are ISO 13485-certified. Bead characteristics and manufacturing conditions help ensure consistency and batch reproducibility making them an ideal choice for commercial supply. If you are in the process of customizing Dynabeads His-Tag Isolation and Pulldown beads in a commercial product or service or need a larger bulk volume of this product, please contact us at oemdynal@lifetech.com or see our Dynabeads OEM page.

Learn more about Dynabeads magnetic beads and KingFisher Sample Purification Systems:
• See our immunoprecipitation selection table, data, and references
• Dynabeads magnetic beads selection guide
• See magnets for Dynabeads magnetic beads separations
• KingFisher automation protocols
• Watch a video about the KingFisher Flex instrument

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Binding Property1.6 mg of a 28 kDa histidine-tagged protein/mL (40 mg) beads
Certifications/ComplianceISO9001 and ISO13485
ColorBrown
Concentration40 mg/mL
Diameter (Metric)1 μm
Final Product TypeHis-Tagged Protein
For Use With (Application)Isolation of His-tagged Recombinant Proteins
For Use With (Equipment)KingFisher Sample Purification System, DynaMag magnets
FormatMagnetic beads
High-throughput CompatibilityYes
Ligand TypeNitrilotriacetic acid (NTA)
MaterialMagnetic Polystyrene
Quantity10 mL
Regulatory StatusRUO
Sample TypeCell Cultures, Tissue Digests, Intact Proteins, Cell Lysates, Cell Extracts
Shelf Life36 months from date of manufacture
Shipping ConditionAmbient Temperature
Sufficient For200 Tests
Surface FunctionalityCobalt-based immobilized metal affinity
TargetHis-tagged proteins
Product LineDynabeads
TypeMagnetic Beads
Unit SizeEach
Contents & Storage
Dynabeads His-Tag Isolation and Pulldown beads are supplied in 20% ethanol. Store at 2°C to 8°C.

Frequently asked questions (FAQs)

My Dynabeads magnetic beads are not pelleting well with the magnet. Do you have any suggestions for me?

Please review the following possibilities for why your Dynabeads magnetic beads are not pelleting:

- The solution is too viscous.
- The beads have formed aggregates because of protein-protein interaction.

Try these suggestions: - Increase separation time (leave tub on magnet for 2-5 minutes)
- Add DNase I to the lysate (~0.01 mg/mL)
- Increase the Tween 20 concentration to ~0.05% of the binding and/or washing buffer.
- Add up to 20 mM beta-merecaptoethanol to the binding and/or wash buffers.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

I have a long double-stranded DNA fragment I would like to isolate. What product do you recommend?

For biotin-labled DNA that is less than 1 kb, we recommend you use Dynabeads M270 Streptavidin and MyOne C1 magnetic beads. We recommend our Dynabeads KilobaseBINDER Kit, which is designed to immobilize long (>1 kb) double-stranded DNA molecules. The KilobaseBINDER reagent consists of M-280 Streptavidin-coupled Dynabeads magnetic beads along with a patented immobilization activator in the binding solution to bind to long, biotinylated DNA molecules for isolation. Please see the following link (https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/napamisc/capture-of-biotinylated-targets/immobilisation-of-long-biotinylated-dna-fragments.html) for more information in regards to long biotinylated DNA fragment isolation.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

Can I use Dynabeads magnetic beads to isolate single-stranded DNA templates?

Yes, Dynabeads magnetic beads can be used to isolate single-stranded DNA. Streptavidin Dynabeads magnetic beads can be used to target biotinylated DNA fragments, followed by denaturation of the double-stranded DNA and removal of the non-biotinylated strand. The streptavidin-coupled Dynabeads magnetic beads will not inhibit any enzymatic activity. This enables further handling and manipulation of the bead-bound DNA directly on the solid phase. Please see the following link (https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/napamisc/capture-of-biotinylated-targets/preparing-single-stranded-dna-templates.html) for more information in regards to single-stranded DNA capture.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

What is the magnetic susceptibility for Dynabeads magnetic beads?

Magnetic susceptibility is a measure of how quickly the beads will migrate to the magnet. This will depend on the iron content and the character of the iron oxide. The magnetic susceptibility given for the Dynabeads magnetic beads is the mass susceptibility, given either as cgs units/g or m^3/kg (the latter being an SI unit). For ferri- and ferromagnetic substances, the magnetic mass susceptibility is dependent upon the magnetic field strength (H), as the magnetization of such substances is not a linear function of H but approaches a saturation value with increasing field. For that reason, the magnetic mass susceptibility of the Dynabeads magnetic beads is determined by a standardized procedure under fixed conditions. The magnetic mass susceptibility given in our catalog is thus the SI unit. Conversion from Gaussian (cgs, emu) units into SI units for magnetic mass susceptibility is achieved by multiplying the Gaussian factor (emu/g or cgs/g) by 4 pi x 10^-3. The resulting unit is also called the rationalized magnetic mass susceptibility, which should be distinguished from the (SI) dimensionless magnetic susceptibility unit. In general, magnetic mass susceptibility is a measure of the force (Fz) influencing an object positioned in a nonhomogenous magnetic field. The magnetic mass susceptibility of the Dynabeads magnetic beads is measured by weighing a sample, and then subjecting the sample to a magnetic field of known strength. The weight (F1) is then measured, and compared to the weight of the sample when the magnetic field is turned off (F0). The susceptibility is then calculated as K x 10^-3 = [(F1-F0) x m x 0.335 x 10^6], where K is the mass susceptibility of the sample of mass m. The susceptibility is then converted to SI units.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

How can I determine coupling efficiency of Dynabeads magnetic beads?

There are different methods to check binding of ligands to the beads, including optical density (OD) measurement, fluorescent labeling, and radioactive labeling.

For OD measurement, you would measure the OD of the ligand before immobilization to the beads and compare it with the ligand concentration that is left in the supernatant after coating. This gives a crude measurement of how much protein has bound to the beads.

Protocol:

1.Set spectrophotometer to the right wavelength. As a blank, use the Coupling Buffer.
2.Measure the absorbance of the Pre-Coupling Solution. A further dilution may be necessary to read the absorbance, depending upon the amount of ligand added.
3.Measure the absorbance of the Post-Coupling Solution. A dilution may be necessary to read the absorbance.
4.Calculate the coupling efficiency, expressed as the % protein uptake, as follows. [(Pre-Coupling Solution x D) - (Post-Coupling Solution x D)] x 100/(Pre-Coupling Solution x D) where D = dilution factor.

For fluorescent labeling, we suggest negatively quantifying the amount of ligand bound by measuring ligand remaining in the coupling supernatant (compared to the original sample), rather than directly measuring the ligands on the beads. Add labeled ligand to the beads, and measure how much ligand is left in the supernatant (not bound to the beads). By comparing this with the total amount added in the first place, you can then calculate how much of the ligand that has been bound to the beads. Keep in mind that the Dynabeads magnetic beads are also autofluorescent, which is why direct measuring of fluorescence of the bead-bound ligands is not recommended, but rather this indirect approach. The label could be, for example, FITC/PE. Some researchers perform a direct approach with success (using a flow cytometer).

Radioactive labeling is the most sensitive method of the three, but it is also the most difficult one. It involves radioactively labeling a portion of the ligand. We use radiolabeled I-125 in tracer amounts and mix it with "cold" ligands in a known ratio before coupling. The absolute quantities for the ligand on the beads should be obtained by measuring the beads in a scintillation (gamma) counter and comparing the cpm with a standard.

Protocol:

1.Take out an appropriate amount of beads and wash the beads in 1 mL of binding buffer.
2.Pipette out desired amount of human IgG in a separate tube.
3.Mix the human IgG with I-125-labeled human IgG (30,000 - 100,000 cpm).
4.Dilute the mixture of human IgG and I-125-labeled human IgG to 100 mL in binding buffer.
5.Incubate for 30 minutes at room temperature and measure the cpm in a scintillation counter.
6.Wash the beads (with coating) four times, and measure cpm again.
The % binding is calculated by using the equation : (cpm after washing/cpm before washing)x100%.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

Citations & References (1)

Citations & References
Abstract
Resolving cadherin interactions and binding cooperativity at the single-molecule level.
Authors:Zhang Y, Sivasankar S, Nelson WJ, Chu S,
Journal:Proc Natl Acad Sci U S A
PubMed ID:19114658
The cadherin family of Ca(2+)-dependent cell adhesion proteins are critical for the morphogenesis and functional organization of tissues in multicellular organisms, but the molecular interactions between cadherins that are at the core of cell-cell adhesion are a matter of considerable debate. A widely-accepted model is that cadherins adhere in 3 ... More