HAAKE™ PolyLab™ QC Modular Torque Rheometer
HAAKE™ PolyLab™ QC Modular Torque Rheometer
HAAKE™ PolyLab™ QC Modular Torque Rheometer
HAAKE™ PolyLab™ QC Modular Torque Rheometer
HAAKE™ PolyLab™ QC Modular Torque Rheometer
Thermo Scientific™
HAAKE™ PolyLab™ QC Modular Torque Rheometer
Confidently meet all your quality control demands with the Thermo Scientific™ HAAKE™ PolyLab™ QC Modular Torque Rheometer.
Have Questions?
Número do catálogoTorque Range
101349640 to 300 Nm
Número do catálogo 10134964
Preço (BRL)
-
Torque Range:
0 to 300 Nm
Confidently meet all your quality control demands with the Thermo Scientific™ HAAKE™ PolyLab™ QC Modular Torque Rheometer. The HAAKE PolyLab QC Modular Torque Rheometer can be connected to an interchangeable mixer, single-screw extruder or conical twin-screw extruder — combining proven technology and state-of-the-art hardware and software with an easy-to-use interface.
You can choose from a bench or floor model of the HAAKE PolyLab QC Modular Torque Rheometer. With an optional rack on the floor model, the instrument can be moved or even stored if needed. Based on the system's backward compatibility, even existing measuring mixers and extruders can be connected to the HAAKE PolyLab QC Modular Torque Rheometer. These devices are automatically recognized by the plug and play Thermo Scientific™ HAAKE™ PolySoft™ Software. This same software also identifies individual limits of the measuring systems, such as torque, speed and temperature and sets alarm values to minimize errors.
The HAAKE PolyLab QC Modular Torque Rheometer Features:
Backward Compatibility
  • Compatible with previous model mixers and extruders
  • Adapts to existing accessories for use
  • Imports previous data for compatibility
Interfaces/Communication
  • CAN open control communication with temperature, motor controller
  • CAN open data communication for pressure and remote signals
  • USB interface to host computer (LAN as option)
Especificações
DescriptionMixers and Extruder System
Measuring Sensor ConnectionCAN open control communication with temperature, motor controller I CAN open data communication for pressure and remote signals I USB interface to host computer (LAN as option)
Temperature Control Zones5
Temperature Reading Channels4
Torque Range0 to 300 Nm
Speed Range0 to 200 rpm
Torque RheometerYes
Unit SizeEach
Frequently asked questions (FAQs)
Why choose the Thermo Scientific HAAKE PolyLab QC system for routine quality control?
The Thermo Scientific HAAKE PolyLab QC system is designed for laboratories performing routine quality control using established test methods. Its compact design, common software platform, and interchangeable processing units support consistent testing while minimizing laboratory space requirements.
Which processing units are compatible with the Thermo Scientific HAAKE PolyLab QC system?
The Thermo Scientific HAAKE PolyLab QC system is compatible with interchangeable laboratory mixers, single-screw laboratory extruders and counter-rotating twin-screw extruders. Using the same drive unit and software platform, laboratories can perform a range of quality control and material characterization tests without changing systems.
Which laboratory is the Thermo Scientific HAAKE PolyLab QC system designed for?
The Thermo Scientific HAAKE PolyLab QC system is designed for laboratories performing routine polymer quality control and standardized material testing. Its compact bench-top design supports day-to-day testing while allowing laboratories to evaluate material processability using compatible laboratory mixers and single-screw extruders.