ImageQuest Software
Thermo Scientific™
ImageQuest Software
Detect, identify and determine the distribution of compounds — all in a single analysis.
Have Questions?
Catalog NumberFor Use With (Equipment)Type
10137985LTQ XL Mass SpectrometersSoftware
Catalog number 10137985
Price (ZAR)
-
For Use With (Equipment):
LTQ XL Mass Spectrometers
Type:
Software
Detect, identify and determine the distribution of compounds—all in a single analysis. Using the power of MSn and the sensitivity of the Thermo Scientific™ LTQ XL mass spectrometer, the Thermo Scientific™ ImageQuest Software saves time while increasing the level of knowledge of the biological system. It enables the visualization of imaging data, creating two-dimensional and three-dimensional maps of analyzed tissue for the optimal display of data.
For small molecule applications, the MALDI LTQ XL with ImageQuest software offers pathologists and pharmaceutical researchers a complete package for the acquisition and presentation of MS images directly from tissue samples.
Intuitive ImageQuest software enables the visualization of imaging data, creating two-dimensional and three-dimensional masps of analyzed tissue for the optimal display of data.
Specifications
DescriptionImageQuest Software
For Use With (Equipment)LTQ XL Mass Spectrometers
TypeSoftware
Unit SizeEach