Interested in a bulk or custom version of this product? Request pricing.

Interleukin 1 Beta

Catalog number:  10139HNAE25

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Interleukin 1, beta (IL1B), a recombinant human protein, is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as immunogens, protein standards, and in other cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the mature form of human IL1B (NP_000567.1) (Ala 117-Ser 269).


Activity:
1.Measured by its binding ability in a functional ELISA.
a. Immobilized recombinant human IL1B at 2 µg/ml (100µl/well) can bind recombinant human IL1R2 with a linear range of 6.25-400 ng/ml.

b. Immobilized human IL1B at 10 µg/ml (100 µl/well) can bind human IL1R1, The EC50 of human IL1R1 is 0.48 µg/ml.

2. Measured in a cell proliferation assay using D10.G4.1 mouse helper T cells. The ED50 for this effect is typically 0.5-5 pg/ml.

Formulation: Lyophilized in 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Cell Cycle & Proliferation, Immunology, Oncology
Expression: E. coli
Form: Lyophilized
Gene Aliases: IL-1, IL1-BETA, IL1F2, IL1B
Gene ID: 3553
Gene Symbol: IL1B
Molecular Weight: 17 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Family: Cytokines & Receptors
Protein Name: interleukin 1, beta
Protein Tag or Fusion: Untagged
Purity: ≥ 90% by SDS-PAGE
Species: Human
Validated Application: Assay Standard

Contents & storage

Includes 5 vials of protein in suspension buffer. Store at -20 °C.

Documents

Manuals & protocols

Recommended products