ARL™ SMS-Omega XRF Instrument
ARL™ SMS-Omega XRF Instrument
ARL™ SMS-Omega XRF Instrument
ARL™ SMS-Omega XRF Instrument
Thermo Scientific™
ARL™ SMS-Omega XRF Instrument
Closely control industrial production to prevent scrap, waste and production rework with an automated XRF system for sample preparation and elemental analysis.
Have Questions?
Catalog NumberType
10144537XRF Instrument
Catalog number 10144537
Price (USD)
-
Type:
XRF Instrument
Keep up with production demands with automated XRF analysis. When different types of material have to be processed in parallel, the Thermo Scientific™ ARL OPTIM'X™ WDXRF Spectrometer with the Thermo Scientific™ ARL™ SMS-Omega XRF Instrument is the solution. Automatic sample preparation and handling with reduced user interaction combined with rapid analysis to provide high-quality results. This innovative solution has a compact footprint of less than 1m², so it can be installed in centralized laboratories or on production shop floors, wherever automated workflow / automated laboratory integration is needed.
This cost-effective system is a unique combination of Thermo Scientific™ XRF and automation technology for automated sample preparation and analysis of one material type.
Main Features
  • Automated operation with reduced routine operator interaction and integrated audit trail functions.
  • Sample handling arm equipped with a suction device (metals version) or a gripper (oxide version) loads samples for analysis.
  • Samples processed under strict identical and reproducible conditions.
  • Fixed circular magazine.
  • XRF functionality comes from the flexible and compact ARL OPTIM'X WDXRF Instrument.
Features of the OPTIM'X WDXRF Instrument
  • WDXRF platform with sequential and/or simultaneous capabilities.
  • Configurable to cover all elements from fluorine (F) to uranium (U) in sequential mode.
  • Fast analysis of solids, liquids and loose powders with superior spectral resolution.
  • No water cooling or gas supply (depending on configuration) required.
  • Closely coupled optics for increased intensity (50% higher than conventional geometry).
Features of the Fixed Circular Magazine
  • Eight positions provides storage of reference standards.
  • Allows occasional manual introduction of prepared samples.
  • One additional position dedicated to filing production samples following analysis.
  • The complete system can also be supplied within a Thermo Scientific™ ARL™ QuantoShelter container for in situ analysis closer to the process.
Specifications
TypeXRF Instrument
Unit SizeEach